Learn More
Biomatik Corporation Human Laminin-5(LN-5)ELISA Kit
Supplier:  Biomatik Corporation EKC34373
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O15230. Target Synonyms: RP11-157P1.6, KIAA1907, laminin alpha 5|laminin alpha-5 chain. Restriction: For Research Use Only.
Catalog No. 50-228-7466