Learn More
Biomatik Corporation Human Mast Cell Tryptase, Mct Elisa Kit
Supplier:  Biomatik Corporation EKC34543
Encompass_Preferred
Human Mast Cell Tryptase, Mct Elisa Kit for detecting Mast Cell Tryptase, Mct in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15661; TPSAB1. Target Synonyms: TPSAB1; TPS1; TPS2; TPSB1; Tryptase alpha/beta-1; Tryptase-1; EC 3.4.21.59; Tryptase I; Tryptase alpha-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7478