Learn More
Biomatik Corporation Human Neurofilament Protein L (Nf-L) Elisa Kit
Supplier:  Biomatik Corporation EKC34781
Encompass_Preferred
Human Neurofilament Protein L (Nf-L) Elisa Kit for detecting Neurofilament, Light Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07196; NEFL. Target Synonyms: NEFL; NF68; NFL; Neurofilament light polypeptide; NF-L; 68 kDa neurofilament protein; Neurofilament triplet L protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7491