Learn More
Biomatik Corporation Human Neuronal Pentraxin-1(NPTX-1) ELISA kit
Supplier:  Biomatik Corporation EKC34798
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q15818. Target Synonyms: DKFZp686J2446, MGC105123, NP1. Restriction: For Research Use Only.
Catalog No. 50-228-7493