Learn More
Biomatik Corporation Human Neuropeptide-Like Protein C4Orf48 (C4Orf48) Elisa Kit
Catalog No. 502287495
Encompass_Preferred
Human Neuropeptide-Like Protein C4Orf48 (C4Orf48) Elisa Kit for detecting Chromosome 4 Open Reading Frame 48 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5BLP8; C4orf48. Target Synonyms: C4orf48; Chr4_55; Neuropeptide-like protein C4orf48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7495 Supplier Biomatik Corporation Supplier No. EKC34804
Add to Cart
Add to Cart
Human Neuropeptide-Like Protein C4Orf48 (C4Orf48) Elisa Kit for detecting Chromosome 4 Open Reading Frame 48 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5BLP8; C4orf48. Target Synonyms: C4orf48; Chr4_55; Neuropeptide-like protein C4orf48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.