Learn More
Biomatik Corporation Human Nuclear matrix protein 22,NMP-22 ELISA Kit
Supplier:  Biomatik Corporation EKC34855
Encompass_Preferred
Sample Type: serum, urine. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q14980. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7497