Learn More
Biomatik Corporation Human Phosphotylinosital 3 kinase,PI3K ELISA Kit
Supplier:  Biomatik Corporation EKC35020
Encompass_Preferred
Sample Type: serum, tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P42336. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7514