Learn More
Biomatik Corporation Human Progesterone Induced Blocking Factor (Pibf) Elisa Kit
Supplier:  Biomatik Corporation EKC35120
Encompass_Preferred
Human Progesterone Induced Blocking Factor (Pibf) Elisa Kit for detecting Progesterone Induced Blocking Factor (Pibf) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q8WXW3; PIBF1. Target Synonyms: PIBF1; C13orf24; PIBF; Progesterone-induced-blocking factor 1; Centrosomal protein of 90 kDa; CEP90. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7524