Learn More
Biomatik Corporation Human Soluble Cluster Of Differentiation 100 (Scd100) Elisa Kit
Supplier:  Biomatik Corporation EKC35592
Encompass_Preferred
Human Soluble Cluster Of Differentiation 100 (Scd100) Elisa Kit for detecting Sema Domain, Immunoglobulin Domain (Ig), Transmembrane Domain (Tm) And Short Cytoplasmic Domain, (Semaphorin) 4D in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92854; SEMA4D. Target Synonyms: A8; BB 18; BB18; CD 100; CD100; CD100 antigen; Coll 4; COLL4; Collapsin 4; Collapsin4; GR3; Leukocyte activation antigen CD100; M sema G; MSEMA; SEM4D_HUMAN; Sema 4d; Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain semaphorin 4D; Sema H; SEMA J; Sema4d; Semacl 2; Semacl2; SemaH; SEMAJ; Semaphorin 4D; Semaphorin C like 2; Semaphorin H; Semaphorin J; Semaphorin-4D; Semaphorin4D; SemaphorinJ; Semcl 2; Semcl2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7544