Learn More
Biomatik Corporation Monkey Complement 3 (C3) Elisa Kit
Supplier:  Biomatik Corporation EKC36146
Encompass_Preferred
Monkey Complement 3 (C3) Elisa Kit for detecting Complement 3(C3) in serum, plasma, cell culture supernates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 4.68-300ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q2UVX4; C3. Target Synonyms: C3Complement C3 [Cleaved into: Complement C3 beta chain; Complement C3 alpha chain; C3a anaphylatoxin; C3-beta-c; C3bc); Acylation stimulating protein; ASP; C3adesArg); Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3dg fragment; Complement C3g fragment; Complement C3d fragment; Complement C3f fragment; Complement C3c alpha' chain fragment 2]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7560