Learn More
Biomatik Corporation Monkey High Sensitivity C-Reactive Protein, Hs-Crp Elisa Kit
Supplier:  Biomatik Corporation EKC36164
Encompass_Preferred
Monkey High Sensitivity C-Reactive Protein, Hs-Crp Elisa Kit for detecting High Sensitivity C-Reactive Protein, Hs-Crp in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7563