Learn More
Biomatik Corporation Mouse ADAM metallopeptidase with thrombospondin type 1 motif, 13 (ADAMTS13) ELISA kit
Supplier:  Biomatik Corporation EKC36255
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q769J6. Target Synonyms: C9orf8, DKFZp434C2322, FLJ42993, MGC118899, MGC118900, TTP, VWFCP, vWF-CP, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13|vWF-cleaving protease|von Wil. Restriction: For Research Use Only.
Catalog No. 50-228-7568