Learn More
Biomatik Corporation Mouse Camp Response Element Binding Protein (Creb) Elisa Kit
Supplier:  Biomatik Corporation EKC36456
Encompass_Preferred
Mouse Camp Response Element Binding Protein (Creb) Elisa Kit for detecting Camp Response Element Binding Protein(Creb) in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q01147; Creb1. Target Synonyms: Creb1; Creb-1Cyclic AMP-responsive element-binding protein 1; CREB-1; cAMP-responsive element-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7578