Learn More
Biomatik Corporation Mouse chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha) (CXCL1) ELISA kit
Supplier:  Biomatik Corporation EKC36536
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P12850. Target Synonyms: FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1, GRO1 oncogene (melanoma growth stimulating activity, alpha)|GRO1 oncogene (melanoma growth-stimulating activity)|MGSA alpha|chemokine (C-X-C motif) ligan. Restriction: For Research Use Only.
Catalog No. 50-228-7581