Learn More
Biomatik Corporation Mouse Choline Acetyltransterase (Chat) Elisa Kit
Supplier:  Biomatik Corporation EKC36546
Encompass_Preferred
Mouse Choline Acetyltransterase (Chat) Elisa Kit for detecting Choline Acetyltransterase(Chat in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q03059; Chat. Target Synonyms: ChatCholine O-acetyltransferase; CHOACTase; ChAT; Choline acetylase; EC 2.3.1.6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7583