Learn More
Biomatik Corporation Mouse Complement 3,C3 ELISA Kit
Supplier:  Biomatik Corporation EKC36625
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 93.75 ng/mL-6000 ng/mL. Sensitivity: 23.44 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01027. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7585