Learn More
Biomatik Corporation Mouse D-Lactate Dehydrogenase, D-Ldh Elisa Kit
Supplier:  Biomatik Corporation EKC36761
Encompass_Preferred
Mouse D-Lactate Dehydrogenase, D-Ldh Elisa Kit for detecting Lactate Dehydrogenase D in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.69-300mU/ml. Sensitivity: 1.17mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q7TNG8; Ldhd. Target Synonyms: Ldhd; Probable D-lactate dehydrogenase; mitochondrial; DLD; Lactate dehydrogenase D; EC 1.1.2.4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7589