Learn More
Biomatik Corporation Mouse Epidermal growth factor,EGF ELISA Kit
Supplier:  Biomatik Corporation EKC36816
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01132. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7591