Learn More
Biomatik Corporation Mouse Immunoglobulin M,IgM ELISA Kit
Supplier:  Biomatik Corporation EKC37087
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 ng/mL-4000 ng/mL. Sensitivity: 62.5 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7600