Learn More
Biomatik Corporation Mouse hemoglobin (Hb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31401
Encompass_Preferred
Mouse Hemoglobin (Hb) Elisa Kit for detecting Hemoglobin (Hb) in serum, plasma, lysate for rbc. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ug/ml. Sensitivity: 0.177ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-7451