Learn More
Biomatik Corporation Mouse N-acetyl-β-D-glucosaminidase,NAG ELISA Kit
Supplier:  Biomatik Corporation EKC37440
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 μIU/mL-1000 μIU/mL. Sensitivity: 3.9 μIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P20060. Target Synonyms: ENC-1AS, N-acetyl-beta-glucosaminidase|hexosaminidase B, Beta-hexosaminidase subunit beta(HEXB). Restriction: For Research Use Only.
Catalog No. 50-228-7614