Learn More
Biomatik Corporation Mouse Retinol-binding protein,RBP ELISA Kit
Supplier:  Biomatik Corporation EKC37744
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 μg/mL-20 μg/mL. Sensitivity: 0.078 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q00724. Target Synonyms: OTTHUMP00000020116|retinol-binding protein 4, interstitial|retinol-binding protein 4, plasma. Restriction: For Research Use Only.
Catalog No. 50-228-7621