Learn More
Biomatik Corporation Mouse thyrotropin-releasing hormone,TRH ELISA KIT
Supplier:  Biomatik Corporation EKC37926
Encompass_Preferred
Sample Type: serum. Species Reactivity: Mus musculus (Mouse). Detection Range: 2.5 pg/mL-160 pg/mL. Sensitivity: 0.625 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q62361. Target Synonyms: MGC125964, MGC125965. Restriction: For Research Use Only.
Catalog No. 50-228-7623