Learn More
Biomatik Corporation Pig Secretory Immunoglobulin A, Siga Elisa Kit
Supplier:  Biomatik Corporation EKC38329
Encompass_Preferred
Pig Secretory Immunoglobulin A, Siga Elisa Kit for detecting Secretory Immunoglobulin A, Siga in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.15-37.5ug/ml. Sensitivity: 0.15ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7637