Learn More
Biomatik Corporation Rabbit Transforming Growth factor β1,TGF-β1 ELISA Kit
Supplier:  Biomatik Corporation EKC38555
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 2.86 ng/mL-50 ng/mL. Sensitivity: 1.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q95215. Target Synonyms: BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1, RGD-containing collagen-associated protein|kerato-epithelin, transforming growth factor-beta1. Restriction: For Research Use Only.
Catalog No. 50-228-7645