Learn More
Biomatik Corporation Rat Cystatin C, Cys-C Elisa Kit
Supplier:  Biomatik Corporation EKC38930
Encompass_Preferred
Rat Cystatin C, Cys-C Elisa Kit for detecting Cystatin C in serum, plasma, homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14841; Cst3. Target Synonyms: Cst3; Cystatin-C; Cystatin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7655