Learn More
Biomatik Corporation Rat Vasoactive Intestinal Peptide,VIP ELISA Kit
Supplier:  Biomatik Corporation EKC39868
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P32649 P01283. Target Synonyms: MGC13587, PHM27. Restriction: For Research Use Only.
Catalog No. 50-228-7683