Learn More
Biomatik Corporation Sheep insulin-like growth factor 1,IGF-1 ELISA Kit
Supplier:  Biomatik Corporation EKC39940
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Ovis aries (Sheep). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P10763. Target Synonyms: IGF1A, IGFI, OTTHUMP00000195084|insulin-like growth factor 1. Restriction: For Research Use Only.
Catalog No. 50-228-7686