Learn More
Biomatik Corporation Human Eosinophil Chemotactic Factor, Ecf Elisa Kit
Supplier:  Biomatik Corporation EKC40036
Encompass_Preferred
Human Eosinophil Chemotactic Factor, Ecf Elisa Kit for detecting Chemokine (C-C Motif) Ligand 11 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51671; CCL11. Target Synonyms: C-C motif chemokine 11; CCL 11; CCL11; CCL11_HUMAN; chemokine (C-C motif) ligand 11; Chemokine C C motif ligand 11; Chemokine CC Motif Ligand 11; Chemokine ligand 11; Eosinophil chemotactic protein; Eotaxin; eotaxin-1; MGC22554; SCYA 11; SCYA11; Small inducible cytokine A11; Small inducible cytokine subfamily A (Cys Cys) member 11; small inducible cytokine subfamily A (Cys-Cys); member 11 (eotaxin); Small inducible cytokine subfamily A member 11; Small-inducible cytokine A11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7693