Learn More
Biomatik Corporation Human Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit
Supplier:  Biomatik Corporation EKC40201
Encompass_Preferred
Human Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit for detecting Bone Gamma-Carboxyglutamate (Gla) Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02818; BGLAP. Target Synonyms: BGLAP; BGP; Bone gamma carboxyglutamate (gla) protein; Bone gamma carboxyglutamate gla protein osteocalcin; Bone gamma carboxyglutamate protein; Bone Gla protein; Gamma carboxyglutamic acid containing protein; Gamma-carboxyglutamic acid-containing protein; OC; OCN; OSTCN_HUMAN; Osteocalcin; OTTHUMP00000016586; PMF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7711