Learn More
Biomatik Corporation Mouse Endothelin 1, Et-1 Elisa Kit
Supplier:  Biomatik Corporation EKC40206
Encompass_Preferred
Mouse Endothelin 1, Et-1 Elisa Kit for detecting Endothelin 1 in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22387; Edn1. Target Synonyms: Edn1Endothelin-1; ET-1; Preproendothelin-1; PPET1) [Cleaved into: Big endothelin-1]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7713