Learn More
Biomatik Corporation Rat Interleukin 5, Il-5 Elisa Kit
Supplier:  Biomatik Corporation EKC40331
Encompass_Preferred
Rat Interleukin 5, Il-5 Elisa Kit for detecting Interleukin 5 (Colony-Stimulating Factor, Eosinophil) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q08125; Il5. Target Synonyms: Il5; Il-5; Interleukin-5; IL-5; B-cell growth factor II; BCGF-II; Cytotoxic T-lymphocyte inducer; Eosinophil differentiation factor; T-cell replacing factor; TRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7719