Learn More
Biomatik Corporation Human Immunoglobulin G (Igg) Elisa Kit
Supplier:  Biomatik Corporation EKC40361
Encompass_Preferred
Human Immunoglobulin G (Igg) Elisa Kit for detecting Immunoglobulin G(Igg) in serum, plasma, cell culture supernates, urine, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.59-150ug/ml. Sensitivity: 0.487ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7726