Learn More
Biomatik Corporation Mouse Neuropeptide Y (Npy) Elisa Kit
Supplier:  Biomatik Corporation EKC40393
Encompass_Preferred
Mouse Neuropeptide Y (Npy) Elisa Kit for detecting Neuropeptide Y in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.87-120pg/ml. Sensitivity: 0.46pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P57774; Npy. Target Synonyms: Npy; Pro-neuropeptide Y [Cleaved into: Neuropeptide Y; Neuropeptide tyrosine; NPY); C-flanking peptide of NPY; CPON)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7730