Learn More
Biomatik Corporation Human Complement 4, C4 Elisa Kit
Supplier:  Biomatik Corporation EKC40500
Encompass_Preferred
Human Complement 4, C4 Elisa Kit for detecting Complement 4, C4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.41-300ng/ml. Sensitivity: 0.41ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P0C0L4; C4A. Target Synonyms: Acidic C4; Acidic complement C4; Basic C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; Chido form; C4; Rodgers from; C4A anaphylatoxin; C4A; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4BD; C4F; C4S; CH; Chido form of C4; CO4; CO4A_HUMAN; Complement C4 A; Complement C4 B; Complement C4 gamma chain; complement C4-A; complement C4-B; complement C4-B-like; complement C4B1a; Complement component 4A (Rodgers blood group); Complement component 4A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7744