Learn More
Biomatik Corporation Human Growth-Regulated Oncogeneα/Melanoma Growth Stimulating Activity, Groα/Mgsa Elisa Kit
Supplier:  Biomatik Corporation EKC40580
Encompass_Preferred
Human Growth-Regulated Oncogeneα/Melanoma Growth Stimulating Activity, Groα/Mgsa Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 1 (Melanoma Growth Stimulating Activity, Alpha) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09341; CXCL1. Target Synonyms: C-X-C motif chemokine 1; Chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity; alpha); chemokine (C-X-C motif) ligand 1; CINC-1; CXCL1; Cytokine-induced neutrophil chemoattractant 1; Fibroblast secretory protein; Fsp; Gro 1; Gro A; Gro; GRO protein; alpha; GRO-alpha (1-73); GRO-alpha (6-73); Gro1; GRO1 oncogene (melanoma growth stimulating activity; alpha); GRO1 oncogene (melanoma growth-stimulating activity). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7757