Learn More
Biomatik Corporation Human Protein C (Pc) Elisa Kit
Supplier:  Biomatik Corporation EKC40710
Encompass_Preferred
Human Protein C (Pc) Elisa Kit for detecting Vitamin K-Dependent Protein C in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04070; PROC. Target Synonyms: Activation peptide; Anticoagulant protein C; APC; Autoprothrombin IIA; Blood coagulation factor XIV; EC 3.4.21.69; PC; proC; PROC_HUMAN; PROC1; Protein C (inactivator of coagulation factors Va and VIIIa); THPH3; THPH4; Vitamin K dependent protein C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7772