Learn More
Biomatik Corporation Rat Interleukin 1 receptor antagonist, IL-1ra ELISA kit
Supplier:  Biomatik Corporation EKC40763
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P25086. Target Synonyms: DIRA, ICIL-1RA, IL-1ra3, IL1F3, IL1RA, IRAP, MGC10430, MVCD4, IL1RN (IL1F3)|OTTHUMP00000203730|intracellular IL-1 receptor antagonist type II|intracellular interleukin-1 receptor antagonist (icIL-1r. Restriction: For Research Use Only.
Catalog No. 50-228-7782