Learn More
Biomatik Corporation Human C4 binding protein,C4BP ELISA Kit
Supplier:  Biomatik Corporation EKC40777
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 39 ng/ml - 2500 ng/ml. Sensitivity: 9.75 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04003. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7784