Learn More
Biomatik Corporation Human Phospholipid Transfer Protein (Pltp) Elisa Kit
Supplier:  Biomatik Corporation EKC40892
Encompass_Preferred
Human Phospholipid Transfer Protein (Pltp) Elisa Kit for detecting Phospholipid Transfer Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.34-150ng/ml. Sensitivity: 0.58ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55058; PLTP. Target Synonyms: BPI fold containing family E; BPIFE; HDLCQ9; High density lipoprotein cholesterol level quantitative trait locus 9; included; Lipid transfer protein II; OD107; Phospholipid transfer protein; Plasma phospholipid transfer protein; Pltp; PLTP_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7794