Learn More
Biomatik Corporation Human Kiaa1199 Elisa Kit
Supplier:  Biomatik Corporation EKC40932
Encompass_Preferred
Human Kiaa1199 Elisa Kit for detecting Kiaa1199 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1-24ng/ml. Sensitivity: 0.4ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8WUJ3; CEMIP. Target Synonyms: CEMIP; KIAA1199; Cell migration-inducing and hyaluronan-binding protein; EC 3.2.1.35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7799