Learn More
Biomatik Corporation Human Interleukin 22 (Il-22) Elisa Kit
Supplier:  Biomatik Corporation EKC40999
Encompass_Preferred
Human Interleukin 22 (Il-22) Elisa Kit for detecting Interleukin 22 in serum, plasma, cell culture supernates, urine, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9GZX6; IL22. Target Synonyms: Cytokine Zcyto18; IL 10 related T cell derived inducible factor; IL 21; IL 22; IL D110; IL TIF; IL-10-related T-cell-derived-inducible factor; IL-22; IL-TIF; IL21; Il22; IL22_HUMAN; ILD110; ILTIF; Interleukin 10 related T cell derived inducible factor; interleukin 21; Interleukin 22; Interleukin-22; MGC79382; MGC79384; TIFa; TIFIL 23; TIFIL23; UNQ3099/PRO10096; zcyto18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7809