Learn More
Biomatik Corporation Rabbit Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit
Supplier:  Biomatik Corporation EKC41098
Encompass_Preferred
Rabbit Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit for detecting Bone Gamma-Carboxyglutamate (Gla) Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P39056; BGLAP. Target Synonyms: BGLAP; Osteocalcin; Bone Gla protein; BGP; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7822