Learn More
Biomatik Corporation Rat Cyclophilin A, Cypa Elisa Kit
Supplier:  Biomatik Corporation EKC41154
Encompass_Preferred
Rat Cyclophilin A, Cypa Elisa Kit for detecting Peptidylprolyl Isomerase A (Cyclophilin A) in serum, plasma, cell lysates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10111; Ppia. Target Synonyms: Ppia; Peptidyl-prolyl cis-trans isomerase A; PPIase A; EC 5.2.1.8; Cyclophilin A; Cyclosporin A-binding protein; Rotamase A; p1B15; p31) [Cleaved into: Peptidyl-prolyl cis-trans isomerase A; N-terminally processed]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7828