Learn More
Biomatik Corporation Human Immunoglobulin D (Igd) Elisa Kit
Supplier:  Biomatik Corporation EKC41166
Encompass_Preferred
Human Immunoglobulin D (Igd) Elisa Kit for detecting Immunoglobulin D(Igd) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.024-100ng/ml. Sensitivity: 0.024ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7830