Learn More
Biomatik Corporation Human Antigen Ki-67 (Ki-67) Elisa Kit
Supplier:  Biomatik Corporation EKC41211
Encompass_Preferred
Human Antigen Ki-67 (Ki-67) Elisa Kit for detecting Antigen Identified By Monoclonal Antibody Ki-67 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P46013; MKI67. Target Synonyms: Antigen identified by monoclonal Ki 67; Antigen identified by monoclonal Ki-67; Antigen KI-67; Antigen KI67; Antigen Ki67; KI67_HUMAN; KIA; Marker of proliferation Ki-67; MIB 1; MIB; MKI67; PPP1R105; Proliferation marker protein Ki-67; Proliferation related Ki 67 antigen; Protein phosphatase 1 regulatory subunit 105; RP11-380J17.2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7836