Learn More
Biomatik Corporation Bovine Alpha-2-Hs-Glycoprotein (Ahsg) Elisa Kit
Supplier:  Biomatik Corporation EKC41347
Encompass_Preferred
Bovine Alpha-2-Hs-Glycoprotein (Ahsg) Elisa Kit for detecting Alpha-2-Hs-Glycoprotein in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 125-8000ng/ml. Sensitivity: 31.25ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12763; AHSG. Target Synonyms: AHSG; FETUAAlpha-2-HS-glycoprotein; Asialofetuin; Fetuin-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7846