Learn More
Biomatik Corporation Human Apolipoprotein C-Iv (Apoc4) Elisa Kit
Supplier:  Biomatik Corporation EKC41363
Encompass_Preferred
Human Apolipoprotein C-Iv (Apoc4) Elisa Kit for detecting Apolipoprotein C-Iv in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P55056; APOC4. Target Synonyms: APO C4; Apo CIV; Apo-CIV; APOC 4; ApoC IV; ApoC-IV; APOC4; APOC4_HUMAN; Apolipoprotein C IV; Apolipoprotein C-IV; Apolipoprotein C4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7848