Learn More
Biomatik Corporation Mouse Soluble Cluster Of Differentiation 40 Ligand, Scd40L Elisa Kit
Supplier:  Biomatik Corporation EKC42221
Encompass_Preferred
Mouse Soluble Cluster Of Differentiation 40 Ligand, Scd40L Elisa Kit for detecting Cd40 Ligand in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P27548; Cd40lg. Target Synonyms: Cd40lg; Cd40l; Tnfsf5; CD40 ligand; CD40-L; T-cell antigen Gp39; TNF-related activation protein; TRAP; Tumor necrosis factor ligand superfamily member 5; CD antigen CD154. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7853