Learn More
Biomatik Corporation Human Glycoprotein Hormones Alpha Chain (Cga) Elisa Kit
Supplier:  Biomatik Corporation EKC42429
Encompass_Preferred
Human Glycoprotein Hormones Alpha Chain (Cga) Elisa Kit for detecting Glycoprotein Hormones, Alpha Polypeptide in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 3-120mIU/ml. Sensitivity: 1mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01215; CGA. Target Synonyms: Anterior pituitary glycoprotein hormones common subunit alpha; CG alpha; CG-alpha; cga; Choriogonadotropin alpha chain; Chorionic Gonadotrophin Alpha; Chorionic Gonadotrophin Alpha Subunit; Chorionic gonadotrophin subunit alpha; chorionic gonadotropin; alpha polypeptide; Follicle-stimulating hormone alpha; Follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; Follitropin alpha; Follitropin alpha chain; FSH Alpha; FSH-alpha; FSHA; GLHA_HUMAN; Glycoprotein hormones alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7884